4-Amino-5-methylpyrimidine 97%

Supplier: Ambeed
Warning

A101952-250MG A101952-1G A101952-5G A101952-100MG
103796-220EA 41 USD
103796-220 103821-914 76780-526 77308-800 77308-802 77599-494
4-Amino-5-methylpyrimidine 97%
4-Amino-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.1300 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909807
CAS Number: 22433-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR