5-Iodocytosine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-iodo-2-pyrimidinol, 4-Amino-5-iodopyrimidin-2(1H)-one

A113479-250MG A113479-1G
103798-238EA 23 USD
103798-238 103824-080 76766-020 77310-896
5-Iodocytosine 98%
5-Iodocytosine

Precaución: for Research Use Only

Formula: C₄H₄IN₃O
MW: 237 g/mol
Melting Pt: 250 °C
Flash Pt: 244.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00023162
CAS Number: 1122-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR