4-Amino-5-chloropyrimidine 95%

Supplier: Ambeed
Warning

A171157-100MG A171157-250MG
103806-600EA 102 USD
103806-600 103844-874 76784-230 76784-232
4-Amino-5-chloropyrimidine 95%
4-Amino-5-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₄ClN₃
Boiling Pt: 253.8 °C (760 mm Hg)
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00233964
CAS Number: 101257-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR