4-Amino-5-chloro-2-ethoxybenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-chloro-2-ethoxybenzoic Acid

A100410-100G A100410-500G
103821-120EA 13 USD
103821-120 77270-156 77270-158
4-Amino-5-chloro-2-ethoxybenzoic acid 97%
4-Amino-5-chloro-2-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
MW: 215.63 g/mol
Boiling Pt: 377.7 °C (760 mm Hg)
Melting Pt: 165…169 °C
Flash Pt: 182.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03840527
CAS Number: 108282-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR