4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A125778-100MG
A125778-250MG
A125778-1G
A125778-25G
103800-720EA
11
USD
103800-720
76822-704
76822-706
77298-084
77885-118
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid 97%
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₈ClNO₃ Boiling Pt: 406.5 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD12923204 CAS Number: 123654-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...