4-Amino-5-bromopyrimidine 98%

Supplier: Ambeed
Warning

A119642-10G A119642-250MG A119642-1G A119642-5G A119642-25G
103799-808EA 25 USD
103799-808 77009-384 77009-386 77297-708 77297-710 77683-074
4-Amino-5-bromopyrimidine 98%
4-Amino-5-bromopyrimidine

Precaución: For research use only.

Formula: C₄H₄BrN₃
MW: 174.00 g/mol
Boiling Pt: 282.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00233962
CAS Number: 1439-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR