4-Amino-5-bromo-2-methylpyridine 97%

Supplier: Ambeed
Warning

A189096-100MG A189096-250MG A189096-25G A189096-1G A189096-5G
103816-156EA 50 USD
103816-156 103846-748 77012-232 77012-234 77012-236 77084-502 77084-504
4-Amino-5-bromo-2-methylpyridine 97%
4-Amino-5-bromo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
Boiling Pt: 276.4 °C (760 mm Hg)
Flash Pt: 120.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368860
CAS Number: 10460-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR