4-Amino-5-bromo-2-chloropyridine 97%

Supplier: Ambeed
Danger

Synonyms: 5-bromo-2-chloropyridin-4-amine, 5-Bromo-2-chloro-4-pyridylamine, 5-Bromo-2-chloro-4-pyridinamine

A139571-100MG A139571-10G A139571-1G A139571-25G A139571-5G
103802-750EA 8 USD
103802-750 77285-664 77285-666 77315-844 77315-846 77315-848
4-Amino-5-bromo-2-chloropyridine 97%
4-Amino-5-bromo-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
Boiling Pt: 315.5 °C (760 mm Hg)
Melting Pt: 127…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09037459
CAS Number: 857730-21-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR