4-Amino-5,6-dichloropyrimidine 95%

Supplier: Ambeed
Warning

A158319-100G A158319-10G A158319-1G
103805-118EA 9 USD
103805-118 103843-328 76768-476 76768-478 77319-186
4-Amino-5,6-dichloropyrimidine 95%
4-Amino-5,6-dichloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₃Cl₂N₃
Boiling Pt: 301.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09837293
CAS Number: 310400-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR