4-Amino-3-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

A246614-25G A246614-10G A246614-1G A246614-5G
103811-720EA 11 USD
103811-720 103851-554 77090-848 77288-896 77304-394 77304-396
4-Amino-3-nitropyridine 98%
4-Amino-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Boiling Pt: 350.4 °C (760 mm Hg)
Melting Pt: 203…207 °C
Flash Pt: 165.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00160304
CAS Number: 1681-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR