2-Nitro-4-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A761375-100G A761375-25G A761375-10G
103840-888EA 6 USD
103840-888 76776-292 77308-272 77573-458
2-Nitro-4-(trifluoromethyl)aniline 98%
2-Nitro-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₂
Boiling Pt: 139.3 °C (760 mm Hg)
Melting Pt: 105…106 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD00007155
CAS Number: 400-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR