4-Amino-m-toluic acid 97%
Supplier: Ambeed
|
Warning
|
A146919-100G
A146919-10G
A146919-1G
A146919-25G
103841-816EA
6
USD
103841-816
77247-136
77247-138
77247-140
77299-554
4-Amino-m-toluic acid 97%
4-Amino-m-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ Boiling Pt: 339.5 °C (760 mm Hg) Melting Pt: 169…171 °C Flash Pt: 159.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007736 CAS Number: 2486-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...