4-Amino-3-methylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

A108465-100MG A108465-10G A108465-250MG A108465-1G A108465-5G
103797-352EA 146 USD
103797-352 103823-132 76765-638 76765-640 76765-642 77309-972 77309-974
4-Amino-3-methylbenzenesulfonamide 97%
4-Amino-3-methylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MW: 186.2300 g/mol
Boiling Pt: 409.3 °C (760 mm Hg)
Flash Pt: 201.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00466840
CAS Number: 53297-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR