4-Amino-3-methoxybenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A105986-100MG
A105986-10G
A105986-250MG
A105986-25G
A105986-1G
A105986-5G
103796-986EA
23
USD
103796-986
103822-736
76970-430
76970-432
76970-434
76970-436
77283-420
77283-422
4-Amino-3-methoxybenzonitrile 98%
4-Amino-3-methoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₈N₂O MW: 148.1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13250177 CAS Number: 177476-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...