4-Amino-3-iodopyridine 98%

Supplier: Ambeed
Danger

Synonyms: 3-Iodopyridin-4-amine, 3-Iodo-4-pyridylamine

A204111-1G A204111-25G A204111-5G A204111-100G A204111-10G
103817-524EA 5 USD
103817-524 103848-188 77086-336 77086-338 77086-340 77152-254 77152-256
4-Amino-3-iodopyridine 98%
4-Amino-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₅IN₂
MW: 220.01 g/mol
Melting Pt: 95…100 °C
Flash Pt: 156.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02663853
CAS Number: 88511-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR