4-Amino-3-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A221505-25G A221505-5G A221505-100G A221505-1G
103818-930EA 8 USD
103818-930 103849-810 77088-404 77088-406 77192-896 77303-604
4-Amino-3-hydroxybenzoic acid 98%
4-Amino-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Boiling Pt: 385.7 °C (760 mm Hg)
Melting Pt: 211…215 °C
Flash Pt: 187.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00017094
CAS Number: 2374-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR