4-Amino-3-fluorophenol 97%

Supplier: Ambeed
Warning

A134488-100G A134488-10G A134488-1G A134488-500G A134488-25G A134488-5G
77285-446EA 119.48 USD
77285-446 77285-448 77285-450 77285-452 77314-772 77314-774
4-Amino-3-fluorophenol 97%
4-Amino-3-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.12 g/mol
Boiling Pt: 263.3 °C (760 mm Hg)
Melting Pt: 135…137 °C
Flash Pt: 113.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00077452
CAS Number: 399-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR