4-Amino-3-chlorophenol hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-chlorophenol Hydrochloride, 2-Chloro-4-hydroxyaniline hydrochloride

A117301-100G A117301-10G A117301-1G A117301-25G A117301-5G
103824-742EA 10 USD
103824-742 77108-324 77108-326 77108-328 77108-330 77311-524
4-Amino-3-chlorophenol hydrochloride 98%
4-Amino-3-chlorophenol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₂NO
MW: 180.04 g/mol
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00143110
CAS Number: 52671-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR