4-Amino-3-chlorophenol 98%

Supplier: Ambeed
Warning

A263642-1G A263642-25G A263642-5G A263642-100G A263642-10G A263642-500G
103812-632EA 5 USD
103812-632 103852-522 77092-162 77092-164 77092-166 77098-866 77098-868 77098-870
4-Amino-3-chlorophenol 98%
4-Amino-3-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 287.3 °C (760 mm Hg)
Melting Pt: 159.5 °C
Flash Pt: 127.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00801116
CAS Number: 17609-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR