4-Amino-3-chlorobenzonitrile 98%

Supplier: Ambeed
Warning

A165380-1G A165380-25G A165380-5G
103805-962EA 5 USD
103805-962 103844-218 77300-780 77300-782 77300-784
4-Amino-3-chlorobenzonitrile 98%
4-Amino-3-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Boiling Pt: 277.8 °C (760 mm Hg)
Melting Pt: 102…105 °C
Flash Pt: 121.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00052917
CAS Number: 21803-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR