4-Amino-3-bromobenzonitrile 97%

Supplier: Ambeed
Warning

A147765-100G A147765-10G A147765-1G A147765-5G A147765-25G
103803-828EA 5 USD
103803-828 103841-936 77137-640 77137-642 77299-600 77299-602 77317-338
4-Amino-3-bromobenzonitrile 97%
4-Amino-3-bromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Boiling Pt: 300.3 °C (760 mm Hg)
Melting Pt: 106…110 °C
Flash Pt: 135.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01861386
CAS Number: 50397-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR