4-Amino-3,5-xylenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-3,5-dimethylphenol
A196120-1G
A196120-25G
A196120-5G
A196120-10G
A196120-250MG
103816-816EA
13
USD
103816-816
103847-430
77085-384
77085-386
77085-388
77192-166
77192-168
4-Amino-3,5-xylenol 98%
4-Amino-3,5-xylenol
Precaución: for Research Use Only
Formula:
C₈H₁₁NO MW: 137.1800 g/mol Boiling Pt: 296.5 °C (760 mm Hg) Melting Pt: 181 °C Flash Pt: 133.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD01741459 CAS Number: 3096-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...