4-Amino-3,5-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-amino-3,5-difluorobenzoicacid

A113913-100MG A113913-250MG A113913-25G A113913-1G A113913-5G
103798-320EA 9 USD
103798-320 103824-162 76810-082 76810-084 76810-086 77310-980 77310-982
4-Amino-3,5-difluorobenzoic acid 97%
4-Amino-3,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.117 g/mol
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD03407440
CAS Number: 500577-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR