4-Amino-3,5-dichlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A126702-100MG A126702-250MG
103800-850EA 364 USD
103800-850 76810-950 76810-952
4-Amino-3,5-dichlorobenzaldehyde 98%
4-Amino-3,5-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
Boiling Pt: 293.5±40.0 °C (760 mm Hg)
Melting Pt: 144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16656998
CAS Number: 62909-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR