4-Amino-3,5-dichloro-6-fluoropyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A102462-100MG A102462-250MG A102462-1G A102462-5G
103796-368EA 149 USD
103796-368 103822-062 76869-196 76869-198 77282-928 77282-930
4-Amino-3,5-dichloro-6-fluoropyridin-2(1H)-one 97%
4-Amino-3,5-dichloro-6-fluoropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₃Cl₂FN₂O
MW: 196.99 g/mol
Boiling Pt: 134.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09751333
CAS Number: 94133-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR