4-Amino-3,5-dibromopyridine 98%
Supplier: Ambeed
|
Warning
|
A428089-25G
A428089-5G
A428089-100G
A428089-10G
A428089-1G
103838-552EA
6
USD
103838-552
76842-224
76842-226
77291-476
77291-478
77291-480
4-Amino-3,5-dibromopyridine 98%
4-Amino-3,5-dibromopyridine
Precaución: for Research Use Only
Formula:
C₅H₄Br₂N₂ Boiling Pt: 292.9 °C (760 mm Hg) Melting Pt: 156…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234048 CAS Number: 84539-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...