4-Amino-2-nitropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Nitropyridin-4-amine

A192777-100MG A192777-1G A192777-250MG
103816-480EA 110 USD
103816-480 103847-090 77012-416 77302-322 77890-181
4-Amino-2-nitropyridine 97%
4-Amino-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Boiling Pt: 382 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04114153
CAS Number: 14916-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR