4-Amino-2-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A285468-250MG A285468-1G A285468-5G A285468-10G
103813-678EA 18 USD
103813-678 103834-812 76827-618 76837-348 76837-350 77683-857
4-Amino-2-methylbenzonitrile 98%
4-Amino-2-methylbenzonitrile

Precaución: For research use only.

Formula: C₈H₈N₂
MW: 132.163 g/mol
Boiling Pt: 298.8 °C (760 mm Hg)
Flash Pt: 134.511 °C
Storage Temperature: Ambient
MDL Number: MFCD09260793
CAS Number: 72115-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR