4-Hydroxy-m-anisidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-methoxyphenol, 4-Aminoguaiacol, 4-Hydroxy-3-methoxyaniline

A332262-25G A332262-5G A332262-100MG A332262-250MG A332262-1G
103825-602EA 11 USD
103825-602 103836-568 76839-564 76839-566 77290-466 77290-468 77306-434
4-Hydroxy-m-anisidine 97%
4-Hydroxy-m-anisidine

Precaución: for Research Use Only

Formula: C₇H₉NO₂
Boiling Pt: 308.1 °C (760 mm Hg)
Flash Pt: 140.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01707809
CAS Number: 52200-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR