4-Amino-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A154283-1G A154283-10G A154283-25G
103804-620EA 13 USD
103804-620 103842-806 77318-512 77592-894 77881-206
4-Amino-2-fluoropyridine 98%
4-Amino-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅FN₂
Boiling Pt: 264 °C (760 mm Hg)
Melting Pt: 83…88 °C
Flash Pt: 113.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095284
CAS Number: 18614-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR