4-Amino-2-fluorobenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A302953-1G
A302953-5G
A302953-250MG
A302953-100G
103814-354EA
18
USD
103814-354
103835-518
76838-206
76838-208
77290-024
77550-832
4-Amino-2-fluorobenzonitrile 98%
4-Amino-2-fluorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₅FN₂ Boiling Pt: 288.2 °C (760 mm Hg) Flash Pt: 128.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07368709 CAS Number: 53312-80-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...