4-Amino-2-chloropyridine 98%

Supplier: Ambeed
Warning

A260832-25G A260832-100G A260832-500G A260832-5G A260832-10G
103852-396EA 6 USD
103852-396 77091-990 77276-544 77276-546 77304-808 77593-626
4-Amino-2-chloropyridine 98%
4-Amino-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00060089
CAS Number: 14432-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR