4-Amino-2-chloropyridin-3-ol 95%

Supplier: Ambeed
Warning

A106210-100MG A106210-250MG A106210-1G
103797-012EA 210 USD
103797-012 76982-428 76982-430 77309-626
4-Amino-2-chloropyridin-3-ol 95%
4-Amino-2-chloropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.559 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16606120
CAS Number: 1227508-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR