4-Amino-2-chlorophenol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-hydroxyaniline

A117642-5G A117642-25G
103824-804EA 13 USD
103824-804 77297-552 77685-040
4-Amino-2-chlorophenol 97%
4-Amino-2-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 292.7 °C (760 mm Hg)
Melting Pt: 150…153 °C
Flash Pt: 130.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00128891
CAS Number: 3964-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR