4-Amino-2-chlorobenzonitrile 98%

Supplier: Ambeed
Warning

A293554-25G A293554-100G A293554-10G A293554-5G
103835-124EA 13 USD
103835-124 76837-744 77141-366 77141-368 77305-636
4-Amino-2-chlorobenzonitrile 98%
4-Amino-2-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
Boiling Pt: 339.5 °C (760 mm Hg)
Melting Pt: 116…118 °C
Flash Pt: 159.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00035926
CAS Number: 20925-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR