4-Amino-2,6-dichlorophenol 97%
Supplier: Ambeed
|
Warning
|
A149988-100G
A149988-10G
A149988-500G
A149988-25G
103808-736EA
144
USD
103808-736
103842-244
77286-096
77286-098
77286-100
77317-756
4-Amino-2,6-dichlorophenol 97%
4-Amino-2,6-dichlorophenol
Precaución: for Research Use Only
Formula:
C₆H₅Cl₂NO Boiling Pt: 303.6 °C (760 mm Hg) Melting Pt: 164…168 °C Flash Pt: 137.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007875 CAS Number: 5930-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...