4-Amino-2,3-difluorobenzoic acid 98%

Supplier: Ambeed
Warning

A187811-1G A187811-5G A187811-100MG A187811-250MG
103816-044EA 22 USD
103816-044 103846-638 77084-350 77084-352 77097-062 77902-022
4-Amino-2,3-difluorobenzoic acid 98%
4-Amino-2,3-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
Boiling Pt: 320.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04114097
CAS Number: 194804-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR