4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine

A229146-25G A229146-5G A229146-100G A229146-10G
103819-448EA 8 USD
103819-448 103850-352 77089-174 77089-176 77288-332 77288-334
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 98%
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO)

Precaución: for Research Use Only

Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
Melting Pt: 37…39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR