4-Amino-2,2,6,6-tetramethylpiperidine 98%
Supplier: Ambeed
|
Warning
|
A611509-25G
A611509-100G
A611509-10G
A611509-500G
103810-480EA
91
USD
103810-480
76844-336
77280-306
77280-308
77280-310
4-Amino-2,2,6,6-tetramethylpiperidine 98%
4-Amino-2,2,6,6-tetramethylpiperidine
Precaución: for Research Use Only
Formula:
C₉H₂₀N₂ Boiling Pt: 188.5 °C (760 mm Hg) Melting Pt: 16…18 °C Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005984 CAS Number: 36768-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...