4-Amino-1-methylpyridin-2-one hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-methylpyridin-2(1H)-one hydrochloride

A119470-100MG A119470-250MG A119470-1G
103799-770EA 66 USD
103799-770 77009-362 77009-364 77311-888
4-Amino-1-methylpyridin-2-one hydrochloride 97%
4-Amino-1-methylpyridin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈N₂O·ClH
MW: 160.601 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20278328
CAS Number: 1404373-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR