4-Amino-1-methylpiperidine 97%

Supplier: Ambeed
Danger

A119589-10G A119589-25G A119589-5G A119589-1G
103799-790EA 9 USD
103799-790 77149-140 77149-142 77297-698 77311-908
4-Amino-1-methylpiperidine 97%
4-Amino-1-methylpiperidine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.1888 g/mol
Boiling Pt: 141 °C (760 mm Hg)
Flash Pt: 39.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01570549
CAS Number: 41838-46-4
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR