4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide 98%
Supplier: Ambeed
|
Warning
|
A116733-100G
A116733-10G
A116733-250MG
A116733-25G
A116733-1G
A116733-5G
103798-788EA
8
USD
103798-788
103824-656
77189-414
77189-416
77189-418
77189-420
77311-450
77311-452
4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide 98%
4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide
Precaución: for Research Use Only
Formula:
C₈H₁₄N₄O Melting Pt: 98…101 °C Flash Pt: 150.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD02927682 CAS Number: 139756-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...