4-Amino-1-methyl-1,2,3-triazole 98%

Supplier: Ambeed
Warning

A317689-100MG A317689-250MG A317689-5G
103825-146EA 294 USD
103825-146 77601-432 77672-628 77881-890
4-Amino-1-methyl-1,2,3-triazole 98%
4-Amino-1-methyl-1,2,3-triazole

Precaución: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₃H₆N₄
Boiling Pt: 272.6 °C (760 mm Hg)
Flash Pt: 118.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09055315
CAS Number: 67545-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR