1H-Pyrrolo[2,3-b]pyridin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1H-pyrrolo[2,3-b]pyridine

A218831-1G A218831-5G A218831-100MG A218831-250MG
103818-750EA 53 USD
103818-750 103849-620 77088-156 77088-158 77192-846 77192-848
1H-Pyrrolo[2,3-b]pyridin-4-amine 97%
1H-Pyrrolo[2,3-b]pyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 348.6±35.0 °C (760 mm Hg)
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD07778366
CAS Number: 74420-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR