4-Amino-1H-indazole 98%

Supplier: Ambeed
Warning

A112610-10G A112610-250MG A112610-1G A112610-5G
103798-054EA 15 USD
103798-054 103823-892 77233-640 77233-642 77297-194 77310-704
4-Amino-1H-indazole 98%
4-Amino-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₇N₃
Boiling Pt: 376.6 °C (760 mm Hg)
Melting Pt: 148…150 °C
Flash Pt: 209.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03305710
CAS Number: 41748-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR