4-Aminoimidazole-5-carboxamide 97%

Supplier: Ambeed
Warning

A144405-10G A144405-500G A144405-25G
103841-476EA 6 USD
103841-476 77272-628 77272-630 77299-376
4-Aminoimidazole-5-carboxamide 97%
4-Aminoimidazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₄H₆N₄O
Boiling Pt: 522.1 °C (760 mm Hg)
Melting Pt: 164…170 °C
Flash Pt: 269.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02181040
CAS Number: 360-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR