5-Iodo-2'-deoxycytidine 95%

Supplier: Ambeed
Warning

A257679-250MG A257679-1G A257679-5G A257679-100G
103812-328EA 10 USD
103812-328 103852-196 76815-944 77091-712 77091-714 77883-702
5-Iodo-2'-deoxycytidine 95%
5-Iodo-2'-deoxycytidine

Precaución: for Research Use Only

Formula: C₉H₁₂IN₃O₄
Boiling Pt: 522.1 °C (760 mm Hg)
Melting Pt: 187…189 °C
Storage Temperature: Freezer
MDL Number: MFCD00038063
CAS Number: 611-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR