4-Acetylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

A353606-25G A353606-100G A353606-10G A353606-5G
103837-104EA 11 USD
103837-104 76840-220 77251-584 77251-586 77306-648
4-Acetylphenylboronic acid 98%
4-Acetylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
Boiling Pt: 354.2 °C (760 mm Hg)
Melting Pt: 240…244 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD01074667
CAS Number: 149104-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR