4-Acetylbenzonitrile 97%

Supplier: Ambeed
Warning

A146457-100G A146457-10G A146457-500G A146457-1G A146457-25G A146457-5G
103803-670EA 5 USD
103803-670 103841-762 77150-274 77150-276 77150-278 77299-524 77317-084 77317-086
4-Acetylbenzonitrile 97%
4-Acetylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00001825
CAS Number: 1443-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR