4-Vinylphenyl acetate 98% (stabilized with MEHQ or TBC) stabilised

Supplier: Ambeed
Warning

Synonyms: Acetic Acid 4-Vinylphenyl Ester, 4-Acetoxystyrene

Stabilization: Stabilized with MEHQ or TBC

A275750-1G A275750-25G A275750-5G A275750-100G A275750-500G A275750-1000G
103813-276EA 5 USD
103813-276 103853-202 77093-060 77093-062 77093-064 77289-538 77289-540 77550-700
4-Vinylphenyl acetate 98% (stabilized with MEHQ or TBC) stabilised
4-Vinylphenyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
Boiling Pt: 260 °C (760 mm Hg)
Melting Pt: 7…8 °C
Flash Pt: 88 °C
Storage Temperature: Freezer
MDL Number: MFCD00075734
CAS Number: 2628-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR